Få mere at vide
3-(Pyridin-2-yl)propiolic Acid, TRC
Artikelnummer. 30422821
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422821 50 mg
30420121 100 mg
30418852 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30422821 Leverandør Toronto Research Chemicals Leverandørnr. TRCP84102850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 858678-71-4
Molekylær formel C8H5NO2
Molekylvægt (g/mol) 147.13
IUPAC navn 3-pyridin-2-ylprop-2-ynoic acid
SMIL OC(=O)C#Cc1ccccn1
Tekniske data
Kemisk navn eller materiale 3-(Pyridin-2-yl)propiolic Acid
Mængde 50 mg
InChI formel InChI=1S/C8H5NO2/c10-8(11)5-4-7-3-1-2-6-9-7/h1-3,6H,(H,10,11)
Formel vægt 147.032