Learn More
3-Propionylpyridine, 98%, Thermo Scientific™
Product Code. 15244388 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15244388 5 g 5g
15234388 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15244388 Supplier Thermo Scientific Acros Supplier No. 454720050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemische Identifikatoren
CAS 1570-48-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
InChI Key VDNKJMUNLKAGAM-UHFFFAOYSA-N
Synonym 3-propionylpyridine, 1-3-pyridyl propan-1-one, ethyl 3-pyridyl ketone, 1-pyridin-3-yl propan-1-one, 3-pyridyl ethyl ketone, 1-3-pyridinyl-1-propanone, 3-propionyl pyridine, 1-pyridin-3-yl-propan-1-one, ethyl pyridin-3-yl ketone, 1-propanone, 1-3-pyridinyl
PubChem CID 74074
IUPAC Name 1-pyridin-3-ylpropan-1-one
SMILES CCC(=O)C1=CN=CC=C1
Spezifikation
Boiling Point 106.0°C (3.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 3-Propionylpyridine
compliance-icons
Produktidentifikator
  • 3-Propionylpyridine, 98%
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.