Få mere at vide
3-(Prop-2-en-1-yl)-1H-indole, TRC
Artikelnummer. 30421184
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421184 50 mg
30419768 100 mg
30420705 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421184 Leverandør Toronto Research Chemicals Leverandørnr. TRCP76884350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16886-09-2
Molekylær formel C11H11N
Molekylvægt (g/mol) 157.21
IUPAC navn 3-prop-2-enyl-1H-indole
SMIL C=CCc1c[nH]c2ccccc12
Tekniske data
Kemisk navn eller materiale 3-(Prop-2-en-1-yl)-1H-indole
Mængde 50 mg
InChI formel InChI=1S/C11H11N/c1-2-5-9-8-12-11-7-4-3-6-10(9)11/h2-4,6-8,12H,1,5H2
Formel vægt 157.0891