Få mere at vide
3-(Prop-2-en-1-yl)-1H-indole, TRC
Artikelnummer. 30419768
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30419768 100 mg
30421184 50 mg
30420705 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30419768 Leverandør Toronto Research Chemicals Leverandørnr. TRCP768843100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16886-09-2
Molekylær formel C11H11N
Molekylvægt (g/mol) 157.21
IUPAC navn 3-prop-2-enyl-1H-indole
SMIL C=CCc1c[nH]c2ccccc12
Tekniske data
Kemisk navn eller materiale 3-(Prop-2-en-1-yl)-1H-indole
Mængde 100 mg
InChI formel InChI=1S/C11H11N/c1-2-5-9-8-12-11-7-4-3-6-10(9)11/h2-4,6-8,12H,1,5H2
Formel vægt 157.0891