Få mere at vide
3-Piperidineacetic Acid-d11, TRC
Artikelnummer. 30415145
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30415145 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415145 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4799421MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Piperidineacetic Acid-d11
CAS 74494-52-3
Molekylær formel C7H2D11NO2
Mængde 1 mg
Synonym 2-(Piperidin-3-yl)acetic Acid-d11, NSC 138735-d11, (Piperidin-3-yl)acetic Acid-d11
InChI formel InChI=1S/C7H13NO2/c9-7(10)4-6-2-1-3-8-5-6/h6,8H,1-5H2,(H,9,10)
SMIL [2H]C([2H])(C(=O)O)C1([2H])C([2H])([2H])NC([2H])([2H])C([2H])([2H])C1([2H])[2H]
Molekylvægt (g/mol) 154.25