missing translation for 'onlineSavingsMsg'
Learn More

3-(Phenylsulfonyl)propylboronic acid diethanolamine ester, Thermo Scientific™

Catalog Number p-7030938
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
C13H20BNO4S
MFCD12546197
3-(Benzenesulfonyl)propylboronic acid diethanolamine ester; 2-[3-(Phenylsulfonyl)propyl]-1,3,6,2-dioxazaborocane
This item is not returnable. View return policy
C13H20BNO4S
MFCD12546197
3-(Benzenesulfonyl)propylboronic acid diethanolamine ester; 2-[3-(Phenylsulfonyl)propyl]-1,3,6,2-dioxazaborocane

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 164°C to 166°C
Quantity 1 g
Formula Weight 297.18
Chemical Name or Material 3-(Phenylsulfonyl)propylboronic acid diethanolamine ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'