missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropionitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7044447
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
645-59-0
C9H9N
131.178
MFCD00001961
ACRWYXSKEHUQDB-UHFFFAOYSA-N
3-phenylpropionitrile, benzenepropanenitrile, hydrocinnamonitrile, 2-phenylethyl cyanide, benzenepropionitrile, 2-cyanoethyl benzene, 3-phenylpropiononitrile, phenylpropionitrile, phenethyl cyanide, hydrocinnamique nitrile
12581
CHEBI:85426
3-phenylpropanenitrile
C1=CC=C(C=C1)CCC#N
This item is not returnable. View return policy
645-59-0
C9H9N
131.178
MFCD00001961
ACRWYXSKEHUQDB-UHFFFAOYSA-N
3-phenylpropionitrile, benzenepropanenitrile, hydrocinnamonitrile, 2-phenylethyl cyanide, benzenepropionitrile, 2-cyanoethyl benzene, 3-phenylpropiononitrile, phenylpropionitrile, phenethyl cyanide, hydrocinnamique nitrile
12581
CHEBI:85426
3-phenylpropanenitrile
C1=CC=C(C=C1)CCC#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -2°C
Density 1.008
Boiling Point 261°C
Flash Point >110°C (230°F)
Refractive Index 1.521
Quantity 10 g
UN Number UN3276
Beilstein 636348
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 3-Phenylpropionitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'