Få mere at vide
3-Phenyl-3-oxetanemethanol, TRC
Artikelnummer. 30411411
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411411 1 g
30421239 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30411411 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3358501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114012-43-0
Molekylær formel C10H12O2
Molekylvægt (g/mol) 164.2
Synonym 3-Hydroxymethyl-3-phenyloxetane, 3-Phenyloxetan-3-ylmethanol
IUPAC navn (3-phenyloxetan-3-yl)methanol
SMIL OCC1(COC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale 3-Phenyl-3-oxetanemethanol
Mængde 1 g
InChI formel InChI=1S/C10H12O2/c11-6-10(7-12-8-10)9-4-2-1-3-5-9/h1-5,11H,6-8H2
Formel vægt 164.0837