Få mere at vide
3-Phenoxybenzaldehyde-d5, TRC
Artikelnummer. 30410477
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30410477 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410477 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29780250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Phenoxybenzaldehyde-d5
CAS 1330277-43-4
Molekylær formel C13H5D5O2
Mængde 50 mg
Synonym m-(Phenyloxy)benzaldehyde-d5, m-Phenoxybenzaldehyde-d5, 5-Phenoxybenzaldehyde-d5
InChI formel InChI=1S/C13H10O2/c14-10-11-5-4-8-13(9-11)15-12-6-2-1-3-7-12/h1-10H/i1D,2D,3D,6D,7D
SMIL [2H]C(C([2H])=C([2H])C([2H])=C1[2H])=C1OC2=CC=CC(C=O)=C2
Molekylvægt (g/mol) 203.25
Formel vægt 203.0995