Få mere at vide
3-Phenanthrol-d9 (Major), TRC
Artikelnummer. 30421605
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30421605 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421605 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2950121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Phenanthrol-d9
CAS 922510-20-1
Molekylær formel C14 2H9 H O
Mængde 1 mg
InChI formel InChI=1S/C14H10O/c15-12-7-8-14-11(9-12)6-5-10-3-1-2-4-13(10)14/h1-9,15H/i1D,2D,3D,4D,5D,6D,7D,8D,9D
SMIL [2H]c1c([2H])c([2H])c2c(c1[2H])c([2H])c([2H])c3c([2H])c(O)c([2H])c([2H])c23
IUPAC navn 1,3,4,5,6,7,8,9,10-nonadeuteriophenanthren-2-ol
Molekylvægt (g/mol) 203.28
Formel vægt 203.1297