Få mere at vide
3-Pentylfuran, TRC
Artikelnummer. 30359647
Skift visning
Mængde:
2.5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359647 10 mg
30356789 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359647 Leverandør Toronto Research Chemicals Leverandørnr. TRCC61779010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Pentylfuran
CAS 6177-84-0
Molekylær formel C9 H14 O
Mængde 10 mg
InChI formel InChI=1S/C9H14O/c1-2-3-4-5-9-6-7-10-8-9/h6-8H,2-5H2,1H3
SMIL CCCCCc1cocc1
IUPAC navn 3-pentylfuran
Molekylvægt (g/mol) 138.207
Formel vægt 138.104