Få mere at vide
3-Pentyl-2,2,3,4,4-d5 Alcohol, CDN
Artikelnummer. 30322923
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322923 0.25 g -
30312530 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322923 Leverandør CDN Leverandørnr. CDND70870.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Pentanol-2,2,3,4,4-D5
CAS 144032-75-7
Molekylær formel C5 D5 H7 O
Mængde 0.25 g
Synonym 3-Pentan-2,2,3,4,4-d5-ol (9 CI), 3-Pentyl-2,2,3,4,4-d5 alcohol, 3-Pentanol-d5, 3-Pentanol-2,2,3,4,4-d5, 3-Pentanol-2,2,3,4,4-D5
InChI formel InChI=1 S/C5H12O/c1-3-5(6)4-2/h5-6 H,3-4H2,1-2H3/i3D2,4D2,5 D
SMIL [2 H]C([2 H])(C)C([2 H])(O)C([2 H])([2 H])C
IUPAC navn 2,2,3,4,4-pentadeuteriopentan-3-ol
Molekylvægt (g/mol) 93.179
Formel vægt 93.1202 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity