Få mere at vide
3-Pentanone-d6, TRC
Artikelnummer. 30409526
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409526 10 mg
30409287 100 mg
30421809 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409526 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27360210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53389-25-6
Molekylær formel C5 2H6 H4 O
Molekylvægt (g/mol) 92.17
Synonym 3-Pentanone-1,1,1,5,5,5,-d6, 3-Pentanone-1,1,1,5,5,5-d6 (5CI), 3-Pentanone-d6, 3-Pentanone-1,1,1,5,5,5-D6, 1,1,1,5,5,5-Hexadeuteropentan-3-one
IUPAC navn 1,1,1,5,5,5-hexadeuteriopentan-3-one
SMIL [2H]C([2H])([2H])CC(=O)CC([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale 3-Pentanone-1,1,1,5,5,5-d6
Mængde 10 mg
InChI formel InChI=1S/C5H10O/c1-3-5(6)4-2/h3-4H2,1-2H3/i1D3,2D3
Formel vægt 92.1108