Få mere at vide
3-Pentanone-2,2,4,4-d4, CDN
Artikelnummer. 30312087
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30312087 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312087 Leverandør CDN Leverandørnr. CDND60181G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Pentanone-2,2,4,4-D4
CAS 6400-97-1
Molekylær formel C5 D4 H6 O
Mængde 1 g
Synonym 3-Pentanone-2,2,4,4-d4 (6 CI,7 CI,8 CI,9 CI), 3-Pentanone-d4, 3-Pentanone-2,2,4,4-D4, 2,2,4,4-Tetradeuteropentan-3-one
InChI formel InChI=1 S/C5H10O/c1-3-5(6)4-2/h3-4H2,1-2H3/i3D2,4D2
SMIL [2 H]C([2 H])(C)C(=O)C([2 H])([2 H])C
IUPAC navn 2,2,4,4-tetradeuteriopentan-3-one
Molekylvægt (g/mol) 90.157
Formel vægt 90.098 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity