Få mere at vide
3-(p-Chlorophenoxy)lactic Acid, TRC
Artikelnummer. 30366066
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366066 50 mg
30351254 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366066 Leverandør Toronto Research Chemicals Leverandørnr. TRCC41486550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(p-Chlorophenoxy)lactic Acid
CAS 7698-98-8
Molekylær formel C9 H9 Cl O4
Mængde 50 mg
Synonym p-Chlorophenoxylactic Acid, 3-(4-Chlorophenoxy)-2-hydroxypropanoic Acid, 3-(4-Chlorophenoxy)-2-hydroxypropionic Acid, NSC 518713
InChI formel InChI=1S/C9H9ClO4/c10-6-1-3-7(4-2-6)14-5-8(11)9(12)13/h1-4,8,11H,5H2,(H,12,13)
SMIL OC(COc1ccc(Cl)cc1)C(=O)O
IUPAC navn 3-(4-chlorophenoxy)-2-hydroxypropanoic acid
Molekylvægt (g/mol) 216.618
Formel vægt 216.019