missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-7Beta-hydroxy-5Beta-cholanoic Acid, TRC

Product Code. 30425449
Change view
Click to view available options
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30425449 100 mg
30410638 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30425449 Supplier Toronto Research Chemicals Supplier No. TRCO856885100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77060-26-5
Molekylær formel C24H38O5
Molekylvægt (g/mol) 390.56
Synonym (5beta,7beta)-7-Hydroxy-3-oxo-cholan-24-oic Acid, 7beta-Hydroxy-3-oxo-5beta-cholan-24-oic Acid, 7beta-Hydroxy-3-oxo-5beta-cholanic Acid, 7beta-Hydroxy-3-keto-5beta-cholanoic Acid, 7beta-Hydroxy-3-oxo-5beta-chol-24-oic Acid, 7beta-Hydroxy-3-oxo-5beta-cholan-24-oic Acid, 7beta-Hydroxy-3-oxo-5beta-cholanic Acid, 7beta-Hydroxy-3-oxo-5beta-cholanoic Acid, 3-Oxo-7beta-hydroxy-5beta-cholanic Acid, 3-Oxo-7beta-hydroxy-5beta-cholanoic Acid, 3-Oxo-7beta-hydroxycholan-24-oic Acid, 3-Oxodeoxycholic Acid
IUPAC navn (4R)-4-[(5R,7S,8R,9S,10S,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Kemisk navn eller materiale 3-Oxo-7beta-hydroxy-5beta-cholanoic Acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C24H38O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-15,17-20,22,26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,17-,18+,19+,20+,22+,23+,24-/m1/s1
Formel vægt 390.277
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.