missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Oxo-7Alpha-hydroxy-5Beta-cholanoic Acid, TRC

Artikelnummer. 30135973
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135973 100 mg
30009329 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30135973 Leverandør Toronto Research Chemicals Leverandørnr. TRCO856875100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4185-00-6
Molekylær formel C24 H38 O4
Molekylvægt (g/mol) 390.56
Synonym Cholan-24-oic acid, 7-hydroxy-3-oxo-, (5β,7α)-, 5β-Cholan-24-oic acid, 7α-hydroxy-3-oxo- (8CI), 5β-Cholanic acid, 7α-hydroxy-3-oxo- (7CI), (5β,7α)-7-Hydroxy-3-oxocholan-24-oic acid, 3-Oxo-7α-hydroxy-5β-cholanic acid, 3-Oxo-7α-hydroxycholan-24-oic acid, 3-Oxochenodeoxycholic acid, 3-keto-7α-Hydroxy-5β-cholanic acid, 7α-Hydroxy-3-oxo-5β-cholan-24-oic acid, 7α-Hydroxy-3-oxo-5β-cholanic acid, 7α-Hydroxy-3-oxo-5β-cholanoic acid
IUPAC navn (4R)-4-[(5R,7R,8R,9S,10S,13R,14S,17R)-7-hydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Kemisk navn eller materiale 3-Oxo-7alpha-hydroxy-5beta-cholanoic Acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C24H38O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-15,17-20,22,26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,17-,18+,19+,20-,22+,23+,24-/m1/s1
Formel vægt 390.277
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.