missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-2,3-dihydro-1H-indazole-5-carboxylic acid, TRC

Product Code. 30422413
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30422413 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422413 Supplier Toronto Research Chemicals Supplier No. TRCO9915285MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Oxo-2,3-dihydro-1H-indazole-5-carboxylic acid
CAS 787580-93-2
Molekylær formel C8H6N2O3
Mængde 5 mg
InChI formel InChI=1S/C8H6N2O3/c11-7-5-3-4(8(12)13)1-2-6(5)9-10-7/h1-3H,(H,12,13)(H2,9,10,11)
SMIL OC(=O)c1ccc2[nH]nc(O)c2c1
IUPAC navn 3-hydroxy-1H-indazole-5-carboxylic acid
Molekylvægt (g/mol) 178.14
Formel vægt 178.0378
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.