missing translation for 'onlineSavingsMsg'
Learn More

3-Oxime Estra-4,9,11-triene-3,17-dione, TRC

Product Code. 30422845
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30422845 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422845 Supplier Toronto Research Chemicals Supplier No. TRCO84685525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Oxime Estra-4,9,11-triene-3,17-dione
CAS 846-56-0
Molekylær formel C18H21NO2
Mængde 25 mg
Synonym (8S,13S,14S,E)-3-(Hydroxyimino)-13-methyl-2,3,7,8,13,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17(6H)-one
InChI formel InChI=1S/C18H21NO2/c1-18-9-8-14-13-5-3-12(19-21)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h8-10,15-16,21H,2-7H2,1H3/b19-12+/t15-,16+,18+/m1/s1
SMIL C[C@@]12[C@](CCC2=O)([H])[C@@]3([H])C(C=C1)=C4C(CC3)=C/C(CC4)=N/O
IUPAC navn (8S,13S,14S,E)-3-(hydroxyimino)-13-methyl-1,2,3,6,7,8,13,14,15,16-decahydro-17H-cyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 283.36
Formel vægt 283.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.