Få mere at vide
3-Oleoylestrone-2,4,16,16-d4, CDN
Artikelnummer. 30313667
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30313667 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313667 Leverandør CDN Leverandørnr. CDND63480.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estrone-2,4,16,16-D4 3-Oleate
CAS 443791-75-1
Molekylær formel C36 D4 H50 O3
Mængde 0.01 g
Synonym Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-[[(9 Z)-1-oxo-9-octadecenyl]oxy]- (9 CI, ACI), 3-[[(9 Z)-1-Oxo-9-octadecenyl]oxy]estra-1,3,5(10)-trien-17-one-2,4,16,16-d4 (ACI), 3-Oleoylestrone-2,4,16,16-d4, Estrone-2,4,16,16-D4 3-oleate, Estrone-2,4,16,16-d4 monooleate, Estrone-2,4,16,16-d4 oleic acid ester, Oleoylestrone-d4
InChI formel InChI=1 S/C36H54O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-35(38)39-29-20-22-30-28(27-29)19-21-32-31(30)25-26-36(2)33(32)23-24-34(36)37/h10-11,20,22,27,31-33 H,3-9,12-19,21,23-26H2,1-2H3/b11-10-/t31-,32-,33+,36+/m1/s1/i20D,24D2,27 D
SMIL [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)[C@@H]3 CCc2c([2 H])c1OC(=O)CCCCCCC\C=C/CCCCCCCC
IUPAC navn [(8 R,9 S,13 S,14 S)-2,4,16,16-tetradeuterio-13-methyl-17-oxo-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-3-yl] (Z)-octadec-9-enoate
Molekylvægt (g/mol) 538.8368
Formel vægt 538.4324 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity