Få mere at vide
3-O-Methyl Colterol-d9, TRC
Artikelnummer. 30470751
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30470751 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470751 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2945725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-O-Methyl Colterol-d9
CAS 1346599-83-4
Renhedsgrad noter HPLC
Molekylær formel C13H12D9NO3
Mængde 5 mg
Synonym α-[[(1,1-Dimethylethyl-d9)amino]methyl]-4-hydroxy-3-methoxybenzenemethanol, N-(tert-Butyl-d9)metarterenol, (+/-)-[(tert-Butyl-d9)amino]methyl]-3-methoxy-4- hydroxybenzyl αlcohol
InChI formel InChI=1S/C13H21NO3/c1-13(2,3)14-8-11(16)9-5-6-10(15)12(7-9)17-4/h5-7,11,14-16H,8H2,1-4H3/i1D3,2D3,3D3
SMIL OC1=C(OC)C=C(C(CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])O)C=C1
IUPAC navn 4-(1-hydroxy-2-((2-(methyl-d3)propan-2-yl-1,1,1,3,3,3-d6)amino)ethyl)-2-methoxyphenol
Molekylvægt (g/mol) 248.37
Formel vægt 248.21
Procent renhed >95