Få mere at vide
3' '-O-Demethyl Ivermectin, TRC
Artikelnummer. 30012919
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012919 5 mg
30010318 1 mg
30004304 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012919 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2928755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 92137-95-6
Molekylær formel C47H72O14
Molekylvægt (g/mol) 861.07
Synonym Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran], avermectin A1a deriv. 3' '-O-Desmethyldihydroavermectin B1a
SMIL C/C([C@H]([C@@H](C)/C=C/C=C1CO[C@@]2([H])[C@]\1(O)[C@@](C(O3)=O)([H])C=C(C)[C@H]2O)O[C@@]4([H])C[C@H](OC)[C@@H](O[C@]5([H])O[C@@H](C)[C@H](O)[C@@H](O)C5)[C@H](C)O4)=C\C[C@]6([H])C[C@@]3([H])C[C@@]7(O[C@@]([C@H](CC)C)([H])[C@@H](C)CC7)O6
Tekniske data
Kemisk navn eller materiale 3' '-O-Demethyl Ivermectin
Mængde 5 mg
InChI formel InChI=1S/C47H72O14/c1-10-24(2)42-27(5)16-17-46(61-42)22-33-19-32(60-46)15-14-26(4)41(58-38-21-36(53-9)43(30(8)56-38)59-37-20-35(48)40(50)29(7)55-37)25(3)12-11-13-31-23-54-44-39(49)28(6)18-34(45(51)57-33)47(31,44)52/h11-14,18,24-25,27,29-30,32-44,48-50,52H,10,15-17,19-23H2,1-9H3/b12-11-,26-14-,31-13?/t24-,25-,27-,29-,30-,32+,33-,34-,35-,36-,37-,38-,39+,40-,41-,42+,43-,44+,46+,47+/m0/s1
Formel vægt 860.4922