Få mere at vide
3-O-Benzyl Estriol, TRC
Artikelnummer. 30338554
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338554 25 mg
30329439 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30338554 Leverandør Toronto Research Chemicals Leverandørnr. TRCB27650025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estriol 3-Benzyl Ether
CAS 18650-87-8
Molekylær formel C25 H30 O3
Mængde 25 mg
Synonym Estriol 3-Benzyl Ether
InChI formel InChI=1S/C25H30O3/c1-25-12-11-20-19-10-8-18(28-15-16-5-3-2-4-6-16)13-17(19)7-9-21(20)22(25)14-23(26)24(25)27/h2-6,8,10,13,20-24,26-27H,7,9,11-12,14-15H2,1H3/t20-,21-,22+,23-,24+,25+/m1/s1
SMIL C[C@]12CC[C@H]3[C@@H](CCc4cc(OCc5ccccc5)ccc34)[C@@H]1C[C@@H](O)[C@@H]2O
IUPAC navn (8R,9S,13S,14S,16R,17R)-13-methyl-3-phenylmethoxy-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-16,17-diol
Molekylvægt (g/mol) 378.5
Formel vægt 378.2195