Få mere at vide
3’-O-Benzyl-(1R)-hydroxy Tapentadol-d5, TRC
Artikelnummer. 30342341
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30342341 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342341 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2799822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3'-O-Benzyl-(1R)-hydroxy Tapentadol-d5
CAS 1246819-60-2
Molekylær formel C21H24D5NO2
Mængde 2.5 mg
Synonym (alphaR)-alpha-[(1S)-2-(Dimethylamino)-1-methylethyl]-alpha-ethyl-3-(phenylmethoxy)benzenemethanol-d5, (2S,3R)-1-(Dimethylamino)-3-[3-(benzyloxy)phenyl)-2-methyl-3-pentanol-d5
InChI formel InChI=1S/C21H29NO2/c1-5-21(23,17(2)15-22(3)4)19-12-9-13-20(14-19)24-16-18-10-7-6-8-11-18/h6-14,17,23H,5,15-16H2,1-4H3/t17-,21-/m1/s1/i1D3,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])[C@@](O)([C@H](C)CN(C)C)c1cccc(OCc2ccccc2)c1
IUPAC navn (3R,4R)-1,1,1,2,2-pentadeuterio-5-(dimethylamino)-4-methyl-3-(3-phenylmethoxyphenyl)pentan-3-ol
Molekylvægt (g/mol) 332.49
Formel vægt 332.2512