Få mere at vide
3-O-Benzyl 16-Epiestriol, TRC
Artikelnummer. 30334191
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334191 5 mg
30347229 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30334191 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2764555MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-O-Benzyl 16-Epiestriol
CAS 33116-58-4
Molekylær formel C25H30O3
Mængde 5 mg
Synonym 3-(Benzyloxy)-estra-1,3,5(10)-triene-16beta,17beta-diol, (16beta,17beta)-3-(Phenylmethoxy)-estra-1,3,5(10)-triene-16,17-diol
InChI formel InChI=1S/C25H30O3/c1-25-12-11-20-19-10-8-18(28-15-16-5-3-2-4-6-16)13-17(19)7-9-21(20)22(25)14-23(26)24(25)27/h2-6,8,10,13,20-24,26-27H,7,9,11-12,14-15H2,1H3/t20-,21-,22+,23+,24+,25+/m1/s1
SMIL C[C@]12CC[C@H]3[C@@H](CCc4cc(OCc5ccccc5)ccc34)[C@@H]1C[C@H](O)[C@@H]2O
IUPAC navn (8R,9S,13S,14S,16S,17R)-13-methyl-3-phenylmethoxy-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-16,17-diol
Molekylvægt (g/mol) 378.5
Formel vægt 378.2195