Få mere at vide
3-O-(Alpha-D-Galactopyranosyl)-D-galactose, TRC
Artikelnummer. 30179634
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179634 5 mg
30179119 10 mg
30179393 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179634 Leverandør Toronto Research Chemicals Leverandørnr. TRCG1547505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13168-24-6
Molekylær formel C12 H22 O11
Molekylvægt (g/mol) 342.3
IUPAC navn (3R,4S,5S,6R)-6-(hydroxymethyl)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-2,3,5-triol
SMIL OC[C@H]1O[C@H](O[C@@H]2[C@@H](O)C(O)O[C@H](CO)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale 3-O-(a-D-Galactopyranosyl)-D-galactose
Mængde 5 mg
InChI formel InChI=1S/C12H22O11/c13-1-3-5(15)7(17)8(18)12(22-3)23-10-6(16)4(2-14)21-11(20)9(10)19/h3-20H,1-2H2/t3-,4-,5+,6+,7+,8-,9-,10+,11?,12-/m1/s1
Formel vægt 342.1162