Få mere at vide
3-O-Acetyl-beta-boswellic Acid, TRC
Artikelnummer. 30011929
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011929 1 mg
30006661 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011929 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1777701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5968-70-7
Molekylær formel C32 H50 O4
Molekylvægt (g/mol) 498.74
Synonym Urs-12-en-23-oic acid, 3-(acetyloxy)-, (3α,4β)-, Urs-12-en-24-oic acid, 3α-hydroxy-, acetate (6CI,8CI), (3α,4β)-3-(Acetyloxy)urs-12-en-23-oic acid, 3-Acetyl-β-boswellic acid, 3-O-Acetyl-β-boswellic acid, 3α-Acetoxyurs-12-en-24-oic acid, 3α-Acetyl-β-boswellic acid, Acetyl-β-boswellic acid, β-Boswellic acid acetate
IUPAC navn (3R,4R,4aR,6aR,6bS,8aR,11R,12S,12aR,14aR,14bR)-3-acetyloxy-4,6a,6b,8a,11,12,14b-heptamethyl-2,3,4a,5,6,7,8,9,10,11,12,12a,14,14a-tetradecahydro-1H-picene-4-carboxylic acid
SMIL C[C@@H]1CC[C@]2(C)CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@@H](OC(=O)C)[C@@](C)([C@@H]5CC[C@@]34C)C(=O)O)[C@@H]2[C@H]1C
Tekniske data
Kemisk navn eller materiale 3-Acetyl-beta-Boswellic acid
Mængde 1 mg
InChI formel InChI=1S/C32H50O4/c1-19-11-14-28(4)17-18-30(6)22(26(28)20(19)2)9-10-23-29(5)15-13-25(36-21(3)33)32(8,27(34)35)24(29)12-16-31(23,30)7/h9,19-20,23-26H,10-18H2,1-8H3,(H,34,35)/t19-,20+,23-,24-,25-,26+,28-,29-,30-,31-,32-/m1/s1
Formel vægt 498.3709