Få mere at vide
3-Nitrobenzyl-d6 Alcohol, CDN
Artikelnummer. 30318488
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318488 0.25 g -
30313625 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318488 Leverandør CDN Leverandørnr. CDND63280.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Nitrobenzyl Alcohol-D6
CAS 1219795-18-2
Molekylær formel C7 D6 H N O3
Mængde 0.25 g
Synonym 3-Nitrobenzyl-d6 Alcohol, 3-Nitrobenzene-d4-methan-d2-ol, (3-Nitrophenyl-2,4,5,6-d4)methanol-d2, 3-Nitrobenzyl alcohol-D6, m-(Hydroxymethyl)nitrobenzene-d6, m-Nitrobenzyl-d6 alcohol
InChI formel InChI=1 S/C7H7NO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4,9 H,5H2/i1D,2 D,3 D,4 D,5D2
SMIL [2 H]c1c([2 H])c(c([2 H])c(c1[2 H])C([2 H])([2 H])O)[N+](=O)[O-]
IUPAC navn dideuterio-(2,3,4,6-tetradeuterio-5-nitrophenyl)methanol
Molekylvægt (g/mol) 159.1724
Formel vægt 159.0803 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity