missing translation for 'onlineSavingsMsg'
Learn More

3-Nitroaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7080897
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
99-09-2
C6H6N2O2
138.13
MFCD00007782
XJCVRTZCHMZPBD-UHFFFAOYSA-N
m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
7423
3-nitroaniline
NC1=CC=CC(=C1)[N+]([O-])=O
This item is not returnable. View return policy
99-09-2
C6H6N2O2
138.13
MFCD00007782
XJCVRTZCHMZPBD-UHFFFAOYSA-N
m-nitroaniline, benzenamine, 3-nitro, 3-nitrophenylamine, nitranilin, 3-aminonitrobenzene, fast orange base r, azobase mna, devol orange r, m-nitrophenylamine, 3-nitrobenzenamine
7423
3-nitroaniline
NC1=CC=CC(=C1)[N+]([O-])=O

3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

Solubility
It is soluble in water 0.1 g/100 ml (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 112°C to 114°C
Density 0.901
Boiling Point 306°C
Quantity 100 g
UN Number UN1661
Beilstein 636962
Merck Index 14,6582
Solubility Information It is soluble in water 0.1g/100 ml (20°C).
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 3-Nitroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'