Få mere at vide
3-Nitroaniline-2,4,5,6-d4, CDN
Artikelnummer. 30327261
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327261 0.5 g 0.5g
30310695 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327261 Leverandør CDN Leverandørnr. CDND54380.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Nitroaniline-2,4,5,6-d4
CAS 115044-52-5
Molekylær formel C6 2H4 H2 N2 O2
Mængde 0.5 g
InChI formel InChI=1 S/C6H6N2O2/c7-5-2-1-3-6(4-5)8(9)10/h1-4 H,7H2/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(N)c([2 H])c(c1[2 H])[N+](=O)[O-]
IUPAC navn 2,3,4,6-tetradeuterio-5-nitroaniline
Molekylvægt (g/mol) 142.15
Formel vægt 142.068 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity