missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitro-L-tyrosine, 98%, Thermo Scientific Chemicals
296.60 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.
Solubility
Soluble in water (partly miscible), 1N hydrochloric acid (50 mg/ml), acidic aqueous solution , ethanol, DMSO, DMF and PBS (pH 7.2) (∼2 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3-Nitro-L-tyrosine is used as an indicator of the formation of peroxynitrite by NO-dependent oxidative damage.
Solubility
Soluble in water (partly miscible), 1N hydrochloric acid (50 mg/ml), acidic aqueous solution , ethanol, DMSO, DMF and PBS (pH 7.2) (∼2 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | ∼230°C (decomposition) |
Beilstein | 2813157 |
Solubility Information | Soluble in water (partly miscible),1N hydrochloric acid (50mg/ml),acidic aqueous solution ,ethanol,DMSO,DMF and PBS (pH 7.2) (∽2mg/ml). |
Formula Weight | 226.19 |
Percent Purity | 98% |
Quantity | 5 g |
Chemical Name or Material | 3-Nitro-L-tyrosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'