missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Catalog Number 15487998
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2822-50-6
C7H5F3N2O3
222.12
MFCD03840207
QXIKEFNBFUHDHK-UHFFFAOYSA-N
3-nitro-4-trifluoromethoxy aniline, 4-trifluoromethoxy-3-nitroaniline, benzenamine,3-nitro-4-trifluoromethoxy, 3-nitro-4-trifluoromethoxy-phenylamine, 5-amino-2-trifluoromethoxy nitrobenzene, pubchem4499, acmc-1cmet, 3-nitro-4-trifluormethoxy-aniline, 3-nitro-4-trifluoromethyloxy aniline, 3-nitro-4-trifluoromethoxy aniline #
613694
3-nitro-4-(trifluoromethoxy)aniline
NC1=CC(=C(OC(F)(F)F)C=C1)[N+]([O-])=O
This item is not returnable. View return policy
2822-50-6
C7H5F3N2O3
222.12
MFCD03840207
QXIKEFNBFUHDHK-UHFFFAOYSA-N
3-nitro-4-trifluoromethoxy aniline, 4-trifluoromethoxy-3-nitroaniline, benzenamine,3-nitro-4-trifluoromethoxy, 3-nitro-4-trifluoromethoxy-phenylamine, 5-amino-2-trifluoromethoxy nitrobenzene, pubchem4499, acmc-1cmet, 3-nitro-4-trifluormethoxy-aniline, 3-nitro-4-trifluoromethyloxy aniline, 3-nitro-4-trifluoromethoxy aniline #
613694
3-nitro-4-(trifluoromethoxy)aniline
NC1=CC(=C(OC(F)(F)F)C=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.543 g/mL
Boiling Point 300°C (760 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.525
Quantity 25 g
Formula Weight 222.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Nitro-4-(trifluoromethoxy)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'