Learn More
3-Nitro-4-hydroxyquinoline, TRC
Product Code. 30469554
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469554 2.5 g 2.5g
30470853 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30469554 Supplier Toronto Research Chemicals Supplier No. TRCN4962102.5G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Nitro-4-hydroxyquinoline
CAS 50332-66-6
Molecular Formula C9 H6 N2 O3
Quantity 2.5 g
InChI Formula InChI=1S/C9H6N2O3/c12-9-6-3-1-2-4-7(6)10-5-8(9)11(13)14/h1-5H,(H,10,12)
SMILES Oc1c(cnc2ccccc12)[N+](=O)[O-]
IUPAC Name 3-nitroquinolin-4-ol
Molecular Weight (g/mol) 190.16
Formula Weight 190.0378