Få mere at vide
Eppendorf™ 3 Neo Single Channel Pipettes
Shop alle Eppendorf produkter
Vægt (metrisk):
74 g
76 g
78 g
80 g
96 g
112 g
Længde (metrisk):
34 mm
40 mm
46 mm
53 mm
55 mm
56 mm
71 mm
115 mm
120 mm
243 mm
Beskrivelse
- Mechanical air-cushion pipette with 2-button operation (one operating element for volume adjustment / pipetting, and one for tip ejection), and customizable settings for easy, ergonomic, and reliable pipetting
- Pipetting performance beyond ISO 8655 requirements when used with original Eppendorf™ pipette tips
- Ergonomic design according to the Eppendorf PhysioCare Concept™ balanced, organic, lightweight, and featuring low pipetting forces
- Spring-loaded tip cones reduce tip attachment and ejection forces, minimizing the risk of strain injuries
- Temporary adjustment to increase accuracy for various liquid types, tip geometries, or altitude without permanently altering the factory calibration
- Removable labeling and marking options with ColorTag pipette marking rings sold separately
- Full autoclavability, high UV and chemical resistance, along with easy disassembly and reassembly, facilitate versatile use and effortless decontamination and cleaning
Greener Choice-anprisninger
- undefined
- Renewable Energy
- Non-Toxic
- Ozone-Safe or Safe for the Ozone Layer
- Recyclable
- Recycled Content
- Source Reduction
Tekniske data
Tekniske data
Certificeringer/compliance | ISO 8655, ACT (US) Label |
Kompatible tips | epT.I.P.S.™ |
Autoklaverbar | Autoclavable |
Farve | White |
Engangs | Reusable |
Til brug med | Pipette Tips |
Greb Type | Ergonomic |
Materiale | Polypropylene |
Antal kanaler | 1 |
Produktlinje | Research |
Vis mere |
Korrektion af produktindhold
Produkttitel
Eppendorf™ 3 Neo Single Channel Pipettes >
Ser du en mulighed for forbedring?Del en indholdskorrektion