Learn More
Eppendorf™ Research 3 Neo Single Channel Pipettes Locking Ring
Description
- Designed for Eppendorf single-channel pipettes
- Ideal when using third-party pipette tips
- Helps deactivate the spring-loaded tip cone feature
- Ensures secure tip attachment and accurate pipetting
- Includes 3 Neo Single Channel Pipettes Locking Rings
Specifications
Specifications
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Locking Clip |
| For Use With (Equipment) | Research plus, Reference 2 Single - Channel Pipettes |
| Quantity | 1 Pk. |