missing translation for 'onlineSavingsMsg'
Få mere at vide

3,N4-Etheno-2’-deoxycytidine, TRC

Artikelnummer. 30135476
Change view
Klicka för att se tillgängliga alternativ
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde
30135476 100 mg
30136304 50 mg
30005607 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30135476 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE677900100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68498-26-0
Molekylær formel C11H13N3O4
Molekylvægt (g/mol) 251.24
Synonym 3,N4-Etheno-dC, 3,N4-Ethenodeoxycytidine, Ethenodeoxycytidine, 6-(2-Deoxy-β-D-erythro-pentofuranosyl)imidazo[1,2-c]pyrimidin-5(6H)-one;
IUPAC navn 6-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)imidazo[1,2-c]pyrimidin-5(6H)-one
SMIL OC[C@H]1O[C@@H](N2C(N3C(C=C2)=NC=C3)=O)C[C@@H]1O

Specifications

Kemisk navn eller materiale 3,N4-Etheno-2'-deoxycytidine
Mængde 100 mg
InChI formel InChI=1S/C11H13N3O4/c15-6-8-7(16)5-10(18-8)14-3-1-9-12-2-4-13(9)11(14)17/h1-4,7-8,10,15-16H,5-6H2/t7-,8+,10+/m0/s1
Formel vægt 287.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.