missing translation for 'onlineSavingsMsg'
Learn More

3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester, 97+%, Thermo Scientific™

Catalog Number 11387539
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
957061-17-5
C15H22BNO4
291.154
MFCD09258775
ATIMPABZLDHDMN-UHFFFAOYSA-N
n-methoxy-n-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-n,o-dimethylhydroxylaminocarbonyl benzeneboronic acid pinacol ester, n-methoxy-n-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb091, 3-n,o-dimethyl hydroxylaminocarbonyl phenyl boronic acid pinacol ester, 3-n,o-dimethyl hydroxylaminocarbonyl phenylboronic acid pinacol ester, 3-n,o-dimethylhydroxylaminocarbonyl phenylboronic acid, pinacol ester, 3-n-methoxy-n-methylcarbamoyl phenylboronic acid pinacol ester, 3-n,o-dimethylhydroxylaminocarbonyl benzeneboronic acid pinacol ester, 97+%
44119763
N-methoxy-N-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)N(C)OC
This item is not returnable. View return policy
957061-17-5
C15H22BNO4
291.154
MFCD09258775
ATIMPABZLDHDMN-UHFFFAOYSA-N
n-methoxy-n-methyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-n,o-dimethylhydroxylaminocarbonyl benzeneboronic acid pinacol ester, n-methoxy-n-methyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, amtb091, 3-n,o-dimethyl hydroxylaminocarbonyl phenyl boronic acid pinacol ester, 3-n,o-dimethyl hydroxylaminocarbonyl phenylboronic acid pinacol ester, 3-n,o-dimethylhydroxylaminocarbonyl phenylboronic acid, pinacol ester, 3-n-methoxy-n-methylcarbamoyl phenylboronic acid pinacol ester, 3-n,o-dimethylhydroxylaminocarbonyl benzeneboronic acid pinacol ester, 97+%
44119763
N-methoxy-N-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)N(C)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 69°C to 72°C
Quantity 1 g
Formula Weight 291.16
Percent Purity ≥97%
Chemical Name or Material 3-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'