missing translation for 'onlineSavingsMsg'
Learn More

3-(N,N-Dicyclohexylsulfamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15441366
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2096331-97-2
C18H28BNO4S
365.30
MFCD20265295
PECBANZNZRTDHD-UHFFFAOYSA-N
3-n,n-dicyclohexylsulfamoyl benzeneboronic acid
73995885
This item is not returnable. View return policy
2096331-97-2
C18H28BNO4S
365.30
MFCD20265295
PECBANZNZRTDHD-UHFFFAOYSA-N
3-n,n-dicyclohexylsulfamoyl benzeneboronic acid
73995885

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 365.29
Percent Purity 97%
Chemical Name or Material 3-(N,N-Dicyclohexylsulfamoyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'