Få mere at vide
3-(N-Methyl-N-pentyl-amino)propionitrile-d3, TRC
Artikelnummer. 30483847
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483847 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483847 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32587910MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(N-Methyl-N-pentyl-amino)-propionitrile-d3
CAS 1185103-36-9
Molekylær formel C9 D3 H15 N2
Mængde 10 mg
Synonym 3-(Pentyl-(trideuteriomethyl)amino)propanenitrile
InChI formel InChI=1S/C9H18N2/c1-3-4-5-8-11(2)9-6-7-10/h3-6,8-9H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])N(CCCCC)CCC#N
IUPAC navn 3-[pentyl(trideuteriomethyl)amino]propanenitrile
Molekylvægt (g/mol) 157.27
Formel vægt 157.1658