Få mere at vide
3-(N-Methyl-N-pentyl-amino)propionic Acid-d3 Hydrochloride, TRC
Artikelnummer. 30469778
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30469778 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469778 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3258785MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-(N-Methyl-N-pentyl-amino)-propionic Acid-d3, Hydrochloride
CAS 1246817-11-7
Molekylær formel C9 2H3 H16 N O2 . Cl H
Mængde 5 mg
InChI formel InChI=1S/C9H19NO2.ClH/c1-3-4-5-7-10(2)8-6-9(11)12;/h3-8H2,1-2H3,(H,11,12);1H/i2D3;
SMIL Cl.[2H]C([2H])([2H])N(CCCCC)CCC(=O)O
IUPAC navn 3-[pentyl(trideuteriomethyl)amino]propanoic acid;hydrochloride
Molekylvægt (g/mol) 212.73
Formel vægt 212.1371