Få mere at vide
3-(N-Acetyl-L-cystein-S-yl) Acetaminophen, TRC
Artikelnummer. 30004302
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30004302 2.5 mg 2.5mg
30004570 10 mg -
30003792 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004302 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1911002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52372-86-8
Molekylær formel C13H16N2O5S
Molekylvægt (g/mol) 312.34
Synonym N-Acetyl-S-[5-(acetylamino)-2-hydroxyphenyl]-L-Cysteine, PCM-M, 3-[(5-Acetamido-2-Hydroxyphenyl)thio]-N-Acetylalanine, Acetaminophen Mercapturate, N-Acetyl-2-(N-Acetyl-L-Cysteinyl)-4-Aminophenol
SMIL OC1=C(SC[C@H](NC(C)=O)C(O)=O)C=C(NC(C)=O)C=C1
Tekniske data
Kemisk navn eller materiale 3-(N-Acetyl-L-cystein-S-yl) Acetaminophen
Renhedsgrad noter HPLC
InChI formel InChI=1S/C13H16N2O5S/c1-7(16)14-9-3-4-11(18)12(5-9)21-6-10(13(19)20)15-8(2)17/h3-5,10,18H,6H2,1-2H3,(H,14,16)(H,15,17)(H,19,20)/t10-/m0/s1
Mængde 2.5 mg
Formel vægt 312.078
Procent renhed >95%
Forsendelsestilstand Dry Pack