missing translation for 'onlineSavingsMsg'
Learn More

3-Methylvaleric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7024359
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
105-43-1
C6H12O2
116.16
MFCD00002729,MFCD02262177
IGIDLTISMCAULB-UHFFFAOYNA-N
3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid
7755
3-methylpentanoic acid
CCC(C)CC(O)=O
This item is not returnable. View return policy
105-43-1
C6H12O2
116.16
MFCD00002729,MFCD02262177
IGIDLTISMCAULB-UHFFFAOYNA-N
3-methylvaleric acid, 3-methyl-pentanoic acid, pentanoic acid, 3-methyl, 3-methyl valeric acid, dl-3-methylvaleric acid, 3-methyl pentanoic acid, sec-butylacetic acid, valeric acid, 3-methyl, beta-methylvaleric acid, 3-methyl-n-valeric acid
7755
3-methylpentanoic acid
CCC(C)CC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -42°C
Density 0.93
Boiling Point 196°C to 198°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.416
Quantity 5 g
UN Number UN3265
Beilstein 1720696
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 3-Methylvaleric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'