Få mere at vide
(±)-3-Methyloctane-d20, CDN
Artikelnummer. 30327332
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327332 0.25 g -
30326596 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327332 Leverandør CDN Leverandørnr. CDND30240.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyloctane-D20
CAS 345909-08-2
Molekylær formel C9 D20
Mængde 0.25 g
Synonym Octane-1,1,1,2,2,3,3,4,4,5,5,6,7,7,8,8,8-d17, 6-(methyl-d3)- (9 CI), 3-Methyloctane-d20, 3-Methyloctane-D20, (±)-3-Methyloctane-d20
InChI formel InChI=1 S/C9H20/c1-4-6-7-8-9(3)5-2/h9H,4-8H2,1-3H3/i1D3,2D3,3D3,4D2,5D2,6D2,7D2,8D2,9 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,6,7,7,8,8,8-heptadecadeuterio-6-(trideuteriomethyl)octane
Molekylvægt (g/mol) 148.3783
Formel vægt 148.282 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity