Få mere at vide
3-Methylfluoranthene, TRC
Artikelnummer. 30133452
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133452 100 mg
30010249 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30133452 Leverandør Toronto Research Chemicals Leverandørnr. TRCM305315100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1706-01-0
Molekylær formel C17H12
Molekylvægt (g/mol) 216.28
Synonym Fluoranthene, 3-methyl-;
IUPAC navn 3-methylfluoranthene
SMIL Cc1ccc2c3ccccc3c4cccc1c24
Tekniske data
Kemisk navn eller materiale 3-Methylfluoranthene
Mængde 100 mg
InChI formel InChI=1S/C17H12/c1-11-9-10-16-14-6-3-2-5-13(14)15-8-4-7-12(11)17(15)16/h2-10H,1H3
Formel vægt 216.0939