Learn More
3-Methylenedihydrofuran-2,5-dione, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 2170-03-8 |
| Molecular Formula | C5H4O3 |
| MDL Number | MFCD00005530 |
| Quantity | 100 g |
| SMILES | C=C(CC1=O)C(O1)=O |
| Molecular Weight (g/mol) | 112.08 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-Methylenedihydrofuran-2,5-dione |