Få mere at vide
3-Methylbutyryl-d9 Chloride, CDN
Artikelnummer. 30308938
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30308938 0.5 g
30320744 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308938 Leverandør CDN Leverandørnr. CDND66080.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methylbutanoyl Chloride-D9
CAS 1219803-46-9
Molekylær formel C5 D9 Cl O
Mængde 0.5 g
Synonym 3-(Methyl-d3)butanoyl-2,2,3,4,4,4-d6 chloride, Isovaleryl chloride-d9, 3-Methylbutyryl chloride-d9, 3-Methylbutyryl-d9 Chloride, 3-Methylbutanoyl chloride-d9, 3-Methylbutanoyl Chloride-D9
InChI formel InChI=1 S/C5H9ClO/c1-4(2)3-5(6)7/h4H,3H2,1-2H3/i1D3,2D3,3D2,4 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(=O)Cl
IUPAC navn 2,2,3,4,4,4-hexadeuterio-3-(trideuteriomethyl)butanoyl chloride
Molekylvægt (g/mol) 129.6328
Formel vægt 129.0907 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity