Få mere at vide
3-Methylbutyric-d9 Acid, CDN
Artikelnummer. 30316590
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316590 0.5 g 0.5g
30309265 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316590 Leverandør CDN Leverandørnr. CDND24540.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isovaleric Acid-d9
CAS 344298-81-3
Molekylær formel C5 2H9 H O2
Mængde 0.5 g
InChI formel InChI=1 S/C5H10O2/c1-4(2)3-5(6)7/h4H,3H2,1-2H3,(H,6,7)/i1D3,2D3,3D2,4 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,4,4,4-hexadeuterio-3-(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 111.19
Formel vægt 111.1246 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity