Få mere at vide
3-Methylbutyraldehyde-2,2-d2, CDN
Artikelnummer. 30321821
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321821 0.5 g
30317097 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321821 Leverandør CDN Leverandørnr. CDND52690.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methylbutyraldehyde-2,2-d2
CAS 352431-47-1
Molekylær formel C5 D2 H8 O
Mængde 0.5 g
InChI formel InChI=1 S/C5H10O/c1-5(2)3-4-6/h4-5 H,3H2,1-2H3/i3D2
SMIL [2 H]C([2 H])(C=O)C(C)C
IUPAC navn 2,2-dideuterio-3-methylbutanal
Molekylvægt (g/mol) 88.145
Formel vægt 88.086 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity