Få mere at vide
3-Methyl-d3-thiopropanal, TRC
Artikelnummer. 30474345
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474345 5 mg
30482553 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474345 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3307725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 136430-27-8
Molekylær formel C4 2H3 H5 S
Molekylvægt (g/mol) 91.19
Synonym Propanal, 3-(methyl-d3-thio)- (9CI), 3-(Methyl-d3-thio)propanal, 3-Methyl-d3-thiopropanal, 3-(Methyl-d3-mercapto)propionaldehyde, Methional-d3, Methional-D3 (S-methyl-D3)
IUPAC navn 4,4,4-trideuteriobutanethial
SMIL [2H]C([2H])([2H])CCC=S
Tekniske data
Kemisk navn eller materiale 3-Methyl-d3-thiopropanal
Mængde 5 mg
InChI formel InChI=1S/C4H8S/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3
Formel vægt 91.0535