Få mere at vide
3-(Methyl-d3-thio)propionaldehyde, CDN
Artikelnummer. 30320113
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320113 0.05 g
30320572 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320113 Leverandør CDN Leverandørnr. CDND78440.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Methyl-d3-thiopropanal
CAS 136430-27-8
Molekylær formel C4 2H3 H5 S
Mængde 0.05 g
InChI formel InChI=1 S/C4H8S/c1-2-3-4-5/h4H,2-3H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])CCC=S
IUPAC navn 4,4,4-trideuteriobutanethial
Molekylvægt (g/mol) 91.19
Formel vægt 91.0535 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity